accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNFFFOCl

CAS RN 126674-98-4

IUPAC Name: 1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride
Synonyms: 1-methyl-3-(trifluoromethyl)pyrazole-4-carbonyl chloride
Molecular Weight (g/mol): 212.56
Molecular Formula: C6H4ClF3N2O
InChi Key: KFKVECZQALNWSR-UHFFFAOYSA-N
SMILES: CN1C=C(C(Cl)=O)C(=N1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results