accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3OBOH3CCH3

CAS RN 126689-01-8

IUPAC Name: 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 168.04
Molecular Formula: C9H17BO2
InChi Key: XGBMQBPLWXTEPM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1CC1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results