accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBr

CAS RN 126930-72-1

IUPAC Name: 1-bromo-4-dodecylbenzene
Synonyms: 1-bromo-4-dodecylbenzene
Molecular Weight (g/mol): 325.33
Molecular Formula: C18H29Br
InChi Key: RFFMRBISTZHCNY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results