accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3CH3CH3H3CCH3(E)(E)(E)(E)

CAS RN 127-47-9

IUPAC Name: (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate
Synonyms: vitamin A acetate 9-cis-retinyl acetate all-trans-retinyl acetate chephasaar brand of retinol acetate novartis brand of retinol acetate davitan A 650 acetate vitamin a all-trans-retinylacetate roche brand of retinol acetate RetiNit
Molecular Weight (g/mol): 328.5
Molecular Formula: C22H32O2
InChi Key: QGNJRVVDBSJHIZ-QHLGVNSISA-N
SMILES: CC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)