accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HCTiClClCl

CAS RN 1270-98-0

IUPAC Name: cyclopenta-2,4-dien-1-ide; trichlorotitaniumylium
Synonyms: cyclopentadienide; trichlorotitaniumylium
Molecular Weight (g/mol): 219.31
Molecular Formula: C5H5Cl3Ti
InChi Key: QOXHZZQZTIGPEV-UHFFFAOYSA-K
SMILES: Cl[Ti+](Cl)Cl.[CH-]1C=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)