accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1271-19-8

IUPAC Name: titanium(2+) bis(cyclopenta-2,4-dien-1-ide) dichloride
Synonyms: titanium(2+) bis(cyclopentadienide) dichloride
Molecular Weight (g/mol): 248.96
Molecular Formula: C10H10Cl2Ti
InChi Key: MKNXBRLZBFVUPV-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Ti++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (5)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (5)
  • (3)
  • (2)