accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HCOsCCCCC

CAS RN 1273-81-0

IUPAC Name: osmocene
Synonyms: osmocene
Molecular Weight (g/mol): 315.38
Molecular Formula: C10H5Os
InChi Key:
SMILES: c1cc[c-]c1.[C-]12[C-]3[C-]4[C-]5[C-]1[Os]2345
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results