accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNHNFFF

CAS RN 127561-18-6

IUPAC Name: 1-[6-(trifluoromethyl)pyridin-2-yl]piperazine
Synonyms: 1-[6-(trifluoromethyl)pyridin-2-yl]piperazine
Molecular Weight (g/mol): 231.22
Molecular Formula: C10H12F3N3
InChi Key: IAOMYUZAXMFANI-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC(=CC=C1)N1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results