accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1277-49-2

FeCH3OH

CAS RN 1277-49-2

IUPAC Name: (1S)-1-(cyclopenta-2,4-dien-1-yl)ethan-1-ol
Synonyms: (1S)-1-(cyclopenta-2,4-dien-1-yl)ethanol
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: KGIBJAFJTDOHBV-LURJTMIESA-N
SMILES: C[C@H](O)C1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results