accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12774-15-1

CHWCo

CAS RN 12774-15-1

IUPAC Name: methylidynetungsten cobalt
Synonyms: methylidynetungsten cobalt
Molecular Weight (g/mol): 255.79
Molecular Formula: CHCoW
InChi Key: IKEHVRMRUVYPGG-UHFFFAOYSA-N
SMILES: [Co].C#[W]
Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results