accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOOH

CAS RN 127958-10-5

IUPAC Name: 2-methyl-4-phenylpyrimidine-5-carboxylic acid
Synonyms: 2-methyl-4-phenylpyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: XKJAQESOTNACHT-UHFFFAOYSA-N
SMILES: CC1=NC=C(C(O)=O)C(=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results