accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 128-08-5

IUPAC Name: 1-bromopyrrolidine-2,5-dione
Synonyms: N-bromosuccinimide N-bromosuccimide NBS succinbromide succinibromimide succinbromimide
Molecular Weight (g/mol): 177.99
Molecular Formula: C4H4BrNO2
InChi Key: PCLIMKBDDGJMGD-UHFFFAOYSA-N
SMILES: BrN1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (7)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (3)
  • (4)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (5)