accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNOO

CAS RN 128-09-6

IUPAC Name: 1-chloropyrrolidine-2,5-dione
Synonyms: chlorosuccinimide succinic N-chloroimide NCS succinchlorimide succinochlorimide
Molecular Weight (g/mol): 133.53
Molecular Formula: C4H4ClNO2
InChi Key: JRNVZBWKYDBUCA-UHFFFAOYSA-N
SMILES: ClN1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)