accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 128-63-2

IUPAC Name: 1,3,6,8-tetrabromopyrene
Synonyms: 1,3,6,8-tetrabromopyrene
Molecular Weight (g/mol): 517.84
Molecular Formula: C16H6Br4
InChi Key: ZKBKRTZIYOKNRG-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=C2C=CC3=C(Br)C=C(Br)C4=CC=C1C2=C34
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results