accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 128-69-8

IUPAC Name: 7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Synonyms: perylenetetracarboxylic dianhydride
Molecular Weight (g/mol): 392.32
Molecular Formula: C24H8O6
InChi Key: CLYVDMAATCIVBF-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results