CAS RN 128196-01-0
CAS RN 128196-01-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Citalopram, TRC
CAS: 128196-01-0 Molecular Formula: C20 H21 F N2 O Molecular Weight (g/mol): 324.39 Synonym: 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, (S)-,(+)-(S)-1-(3-(Dimethylamino)propyl)-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile,(+)-(S)-Citalopram,(+)-Citalopram,(S)-Citalopram,Escitalopram,S-(+)-Citalopram,Seroplex;5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, (1S)- IUPAC Name: (1S)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile SMILES: CN(C)CCC[C@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3