accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SFFF

CAS RN 128273-56-3

IUPAC Name: 1-{4-[(trifluoromethyl)sulfanyl]phenyl}methanamine
Synonyms: 1-{4-[(trifluoromethyl)sulfanyl]phenyl}methanamine
Molecular Weight (g/mol): 207.21
Molecular Formula: C8H8F3NS
InChi Key: LACURGWEZCFLBO-UHFFFAOYSA-N
SMILES: NCC1=CC=C(SC(F)(F)F)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results