accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOOOOBr

CAS RN 128611-93-8

IUPAC Name: (2-bromophenyl)methyl 2,5-dioxopyrrolidin-1-yl carbonate
Synonyms: (2-bromophenyl)methyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular Weight (g/mol): 328.12
Molecular Formula: C12H10BrNO5
InChi Key: SZDNRTVADOCWKU-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1COC(=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results