accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 128625-52-5

IUPAC Name: (1H-1,2,3-benzotriazol-1-yloxy)tris(pyrrolidin-1-yl)phosphanium
Synonyms: PyBOP
Molecular Weight (g/mol): 375.44
Molecular Formula: C18H28N6OP
InChi Key: WGNZRLMOMHJUSP-UHFFFAOYSA-N
SMILES: C1CCN(C1)[P+](ON1N=NC2=CC=CC=C12)(N1CCCC1)N1CCCC1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)