CAS RN 128733-85-7
IUPAC Name:
(5-tert-butyl-2-methoxyphenyl)boronic acid
Synonyms:
5-tert-butyl-2-methoxyphenylboronic acid
Molecular Weight (g/mol):
208.06
Molecular Formula:
C11H17BO3
InChi Key:
LXFOJKCQSAYVMF-UHFFFAOYSA-N
SMILES:
COC1=C(C=C(C=C1)C(C)(C)C)B(O)O
1
–
1
of
1
results
1
5-tert-Butyl-2-methoxybenzeneboronic acid, 98+%, Thermo Scientific Chemicals
CAS: 128733-85-7 Molecular Formula: C11H17BO3 Molecular Weight (g/mol): 208.064 MDL Number: MFCD06201034 InChI Key: LXFOJKCQSAYVMF-UHFFFAOYSA-N Synonym: 5-tert-butyl-2-methoxyphenyl boronic acid,5-t-butyl-2-methoxyphenylboronic acid,5-tert-butyl-2-methoxybenzeneboronic acid,2-borono-4-tert-butyl anisole,acmc-1c4zr,2-methoxy-5-tert-butylphenylboronic acid,5-tert-butyl-2-methoxyphenyl boronicacid,boronic acid, b-5-1,1-dimethylethyl-2-methoxyphenyl PubChem CID: 14740579 IUPAC Name: (5-tert-butyl-2-methoxyphenyl)boronic acid SMILES: B(C1=C(C=CC(=C1)C(C)(C)C)OC)(O)O