accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 129-00-0

IUPAC Name: pyrene
Synonyms: pyrene β-pyrene benzo(def)phenanthrene
Molecular Weight (g/mol): 202.26
Molecular Formula: C16H10
InChi Key: BBEAQIROQSPTKN-UHFFFAOYSA-N
SMILES: C1=CC2=CC=C3C=CC=C4C=CC(=C1)C2=C34
Molecular Weight (g/mol) 
  • (6)
  • (11)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
Concentration 
  • (4)
  • (4)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (8)
  • (1)
  • (3)
  • (2)
  • (6)
  • (11)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)