accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 129-73-7

H3CNCH3NCH3H3C

CAS RN 129-73-7

IUPAC Name: 4-{[4-(dimethylamino)phenyl](phenyl)methyl}-N,N-dimethylaniline
Synonyms: 4-{[4-(dimethylamino)phenyl](phenyl)methyl}-N,N-dimethylaniline
Molecular Weight (g/mol): 330.48
Molecular Formula: C23H26N2
InChi Key: WZKXBGJNNCGHIC-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)