CAS RN 1290541-46-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2
3-(Phenylmethyl)-2-[(1E)-2-(3-pyridinyl)ethenyl]-4(3H)-quinazolinone, TRC
CAS: 1290541-46-6 Molecular Formula: C12 H17 N3 O6 Molecular Weight (g/mol): 299.28 Synonym: BO 2 IUPAC Name: ethyl 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidine-5-carboxylate SMILES: CCOC(=O)C1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N=C1N