accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1291-32-3

ClZrCl

CAS RN 1291-32-3

IUPAC Name: dichlorobis(cyclopenta-2,4-dien-1-yl)zirconium
Synonyms: zirconocene dichloride
Molecular Weight (g/mol): 292.31
Molecular Formula: C10H10Cl2Zr
InChi Key: QRUYYSPCOGSZGQ-UHFFFAOYSA-L
SMILES: Cl[Zr](Cl)(C1C=CC=C1)C1C=CC=C1
Physical Form 
  • (3)
  • (7)
Quantity 
  • (1)
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (7)
Percent Purity 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (7)
  • (3)
  • (7)
  • (1)
  • (3)
  • (3)
  • (3)
  • (7)
  • (4)
  • (3)