accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 129368-70-3

H3CH3CCH3SiH3CCH3OBr

CAS RN 129368-70-3

IUPAC Name: [(6-bromohexyl)oxy](tert-butyl)dimethylsilane
Synonyms: [(6-bromohexyl)oxy](tert-butyl)dimethylsilane
Molecular Weight (g/mol): 295.34
Molecular Formula: C12H27BrOSi
InChi Key: PBKXRKYUUXKNSL-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OCCCCCCBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results