accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(S)(S)(R)(R)SOFFFFFHO(S)(S)OH

CAS RN 129453-61-8

IUPAC Name: (1S,3aS,3bR,4R,9bS,11aS)-11a-methyl-4-[9-(4,4,5,5,5-pentafluoropentanesulfinyl)nonyl]-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Synonyms: fulvestrant
Molecular Weight (g/mol): 606.78
Molecular Formula: C32H47F5O3S
InChi Key: VWUXBMIQPBEWFH-WCCTWKNTSA-N
SMILES: C[C@]12CC[C@H]3[C@H]([C@@H]1CC[C@@H]2O)[C@H](CCCCCCCCCS(=O)CCCC(F)(F)C(F)(F)F)CC1=CC(O)=CC=C31
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results