accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 129499-78-1

IUPAC Name: (2R)-2-[(1S)-1,2-dihydroxyethyl]-5-hydroxy-4-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2,3-dihydrofuran-3-one
Synonyms: ascorbyl glucoside
Molecular Weight (g/mol): 338.27
Molecular Formula: C12H18O11
InChi Key: YEERNPPBIMJBHH-DCWMUDTNSA-N
SMILES: OC[C@H](O)[C@H]1OC(O)=C(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results