accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SFFFFFFSOOO

CAS RN 129946-88-9

IUPAC Name: 8-(trifluoromethyl)-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaen-8-ium trifluoromethanesulfonate
Synonyms: 8-(trifluoromethyl)-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaen-8-ium triflate
Molecular Weight (g/mol): 402.33
Molecular Formula: C14H8F6O3S2
InChi Key: QXXHXTRTGZBOGD-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.FC(F)(F)[S+]1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)