accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO

CAS RN 130-15-4

IUPAC Name: 1,4-dihydronaphthalene-1,4-dione
Synonyms: naphthoquinone 1,4-naphthylquinone succivil usaf CY-10 1,4-naftochinon α-naphthoquinone rcra waste number U166
Molecular Weight (g/mol): 158.16
Molecular Formula: C10H6O2
InChi Key: FRASJONUBLZVQX-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)