accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 130-16-5

OHClN

CAS RN 130-16-5

IUPAC Name: 5-chloroquinolin-8-ol
Synonyms: 5-chloro-8-hydroxyquinoline cloxyquin 氯羟喹 cloxiquinum 8-quinolinol,5-chloro- chloroxychinolin dermofongin cloxiquine cloxiquina 5-chlor-8-chinolinol
Molecular Weight (g/mol): 179.6
Molecular Formula: C9H6ClNO
InChi Key: CTQMJYWDVABFRZ-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)