accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 130-95-0

IUPAC Name: {5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(6-methoxyquinolin-4-yl)methanol
Synonyms: quinine quinine salicylate (-)-quinine quinidine, dihydrate biioquinol 奎宁
Molecular Weight (g/mol): 324.42
Molecular Formula: C20H24N2O2
InChi Key: LOUPRKONTZGTKE-UHFFFAOYNA-N
SMILES: COC1=CC=C2N=CC=C(C(O)C3CC4CCN3CC4C=C)C2=C1
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (6)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (6)
  • (3)