accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SONHBrOFFFBrNFFF

CAS RN 130000-40-7

IUPAC Name: N-[2,6-dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxamide
Synonyms: thifluzamide
Molecular Weight (g/mol): 528.06
Molecular Formula: C13H6Br2F6N2O2S
InChi Key: WOSNCVAPUOFXEH-UHFFFAOYSA-N
SMILES: CC1=NC(=C(S1)C(=O)NC1=C(Br)C=C(OC(F)(F)F)C=C1Br)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results