accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13007-90-4

NiPPOOCC

CAS RN 13007-90-4

IUPAC Name: bis(methanidylidyneoxidanium) bis(triphenylphosphanium) nickel
Synonyms: bis(carbon monoxide) bis(triphenylphosphanium) nickel
Molecular Weight (g/mol): 641.31
Molecular Formula: C38H32NiO2P2
InChi Key: SLFKPACCQUVAPG-UHFFFAOYSA-P
SMILES: [Ni].[C-]#[O+].[C-]#[O+].C1=CC=C(C=C1)[PH+](C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)[PH+](C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results