accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OO(R)H3CCH3OO

CAS RN 130165-76-3

IUPAC Name: (3R)-3-(methoxycarbonyl)-5-methylhexanoate
Synonyms: (3R)-3-(methoxycarbonyl)-5-methylhexanoate
Molecular Weight (g/mol): 187.22
Molecular Formula: C9H15O4
InChi Key: AJNNFMRCMKGYHA-SSDOTTSWSA-M
SMILES: COC(=O)[C@H](CC(C)C)CC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)