accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1302-78-9

IUPAC Name: tetrasilicon(4+) dialuminium(3+) hydrate undecaoxidandiide
Synonyms: tetrasilicon(4+) dialuminium(3+) hydrate undecaoxidandiide
Molecular Weight (g/mol): 360.31
Molecular Formula: Al2H2O12Si4
InChi Key: KMDMICAGCNGGEB-UHFFFAOYSA-N
SMILES: O.[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Si+4].[Si+4].[Si+4].[Si+4]
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
Grade 
  • (4)
  • (2)
Material 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)