accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

AlAlSSS

CAS RN 1302-81-4

IUPAC Name: dialuminium(3+) trisulfanediide
Synonyms: aluminium sulfide
Molecular Weight (g/mol): 150.14
Molecular Formula: Al2S3
InChi Key: COOGPNLGKIHLSK-UHFFFAOYSA-N
SMILES: [Al+3].[Al+3].[S--].[S--].[S--]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results