CAS RN 13023-73-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dihydroxyphenylglycolaldehyde (Dopegal), TRC
CAS: 13023-73-9 Molecular Formula: C8 H8 O4 Molecular Weight (g/mol): 168.15 Synonym: 3,4-Dihydroxyphenylglycolic Aldehyde; IUPAC Name: 2-(3,4-dihydroxyphenyl)-2-hydroxyacetaldehyde SMILES: OC(C=O)c1ccc(O)c(O)c1