CAS RN 130232-51-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ciprofibrate Methyl Ester, TRC
CAS: 130232-51-8 Molecular Formula: C14H16Cl2O3 Molecular Weight (g/mol): 303.18 Synonym: 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methyl-propanoic Acid Methyl Ester,(±)-2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methyl-propanoic Acid Methyl Ester; IUPAC Name: methyl 2-[4-(2,2-dichlorocyclopropyl)phenoxy]-2-methylpropanoate SMILES: COC(=O)C(C)(C)Oc1ccc(cc1)C2CC2(Cl)Cl