accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13026-23-8

IUPAC Name: (2Z)-3-{[1,1'-biphenyl]-4-yl}prop-2-enoic acid
Synonyms: (2Z)-3-{[1,1'-biphenyl]-4-yl}prop-2-enoic acid
Molecular Weight (g/mol): 224.26
Molecular Formula: C15H12O2
InChi Key: DMJDEZUEYXVYNO-FLIBITNWSA-N
SMILES: OC(=O)\C=C/C1=CC=C(C=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results