accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13031-41-9

CH3OON

CAS RN 13031-41-9

IUPAC Name: 4-cyanophenyl acetate
Synonyms: 4-cyanophenyl acetate
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: CJGXWABHYYJNJH-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results