CAS RN 1304-76-3
Bismuth(III) oxide, 99.9%, (trace metal basis)
CAS: 1304-76-3 Molecular Formula: Bi2O3 Molecular Weight (g/mol): 465.96 MDL Number: MFCD00003462 InChI Key: TYIXMATWDRGMPF-UHFFFAOYSA-N Synonym: Bismuth trioxide IUPAC Name: dibismuth(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Bi+3].[Bi+3]
Bismuth(III) oxide, 99.975% (metals basis)
CAS: 1304-76-3 Molecular Formula: Bi2O3 Molecular Weight (g/mol): 465.96 MDL Number: MFCD00003462 InChI Key: TYIXMATWDRGMPF-UHFFFAOYSA-N Synonym: Bismuth salicylate, basic,Salicylic acid bismuth basic salt IUPAC Name: dibismuth(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Bi+3].[Bi+3]
Bismuth(III) oxide, 99%
CAS: 1304-76-3 Molecular Formula: Bi2O3 Molecular Weight (g/mol): 465.96 MDL Number: MFCD00003462 InChI Key: TYIXMATWDRGMPF-UHFFFAOYSA-N IUPAC Name: dibismuth(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Bi+3].[Bi+3]
Bismuth(III) oxide, Puratronic™, 99.999% (metals basis)
CAS: 1304-76-3 Molecular Formula: Bi2O3 Molecular Weight (g/mol): 465.96 MDL Number: MFCD00003462 InChI Key: TYIXMATWDRGMPF-UHFFFAOYSA-N IUPAC Name: dibismuth(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Bi+3].[Bi+3]
Bismuth(III) oxide, Puratronic™, 99.9995% (metals basis)
CAS: 1304-76-3 | Bi{2}O{3}
Bismuth(III) oxide, typically 99.99% (metals basis)
CAS: 1304-76-3 Molecular Formula: Bi2O3 Molecular Weight (g/mol): 465.96 MDL Number: MFCD00003462 InChI Key: TYIXMATWDRGMPF-UHFFFAOYSA-N IUPAC Name: dibismuth(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Bi+3].[Bi+3]
Bismuth(III) oxide, nanopowder, 99.9%
CAS: 1304-76-3 Molecular Formula: Bi2O3 Molecular Weight (g/mol): 465.96 MDL Number: MFCD00003462 InChI Key: TYIXMATWDRGMPF-UHFFFAOYSA-N IUPAC Name: dibismuth(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Bi+3].[Bi+3]
Bismuth(III) oxide, Puratronic™, 99.9995% (metals basis)
CAS: 1304-76-3 Molecular Formula: Bi2O3 Molecular Weight (g/mol): 465.96 MDL Number: MFCD00003462 InChI Key: TYIXMATWDRGMPF-UHFFFAOYSA-N IUPAC Name: dibismuth(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Bi+3].[Bi+3]
Bismuth(III) Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Bismuth(III) Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Bismuth(III) Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Bismuth(III) oxide, NanoArc™, 99.5+%
CAS: 1304-76-3 Molecular Formula: Bi2O3 Molecular Weight (g/mol): 465.96 MDL Number: MFCD00003462 InChI Key: TYIXMATWDRGMPF-UHFFFAOYSA-N IUPAC Name: dibismuth(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Bi+3].[Bi+3]