accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1304-85-4

ONOOBiONOOONOO

CAS RN 1304-85-4

IUPAC Name: bis(nitrooxy)bismuthanyl nitrate
Synonyms: bismuth subnitrate
Molecular Weight (g/mol): 394.99
Molecular Formula: BiN3O9
InChi Key: PPNKDDZCLDMRHS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)O[Bi](O[N+]([O-])=O)O[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)