accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOH3C

CAS RN 13041-60-6

IUPAC Name: methyl 3-methoxynaphthalene-2-carboxylate
Synonyms: methyl 3-methoxynaphthalene-2-carboxylate
Molecular Weight (g/mol): 216.24
Molecular Formula: C13H12O3
InChi Key: MPLSAOVNAXJHHI-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(OC)C=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)