accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHO

CAS RN 130423-83-5

IUPAC Name: 5-(2-phenylethynyl)furan-2-carboxylic acid
Synonyms: 5-(2-phenylethynyl)furan-2-carboxylic acid
Molecular Weight (g/mol): 212.2
Molecular Formula: C13H8O3
InChi Key: HFYACYZCOUYIFR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O1)C#CC1=CC=CC=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results