accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1306-06-5

IUPAC Name: pentacalcium hydroxide triphosphate
Synonyms: hydroxyapatite calcium hydroxyapatite
Molecular Weight (g/mol): 502.31
Molecular Formula: Ca5HO13P3
InChi Key: XYJRXVWERLGGKC-UHFFFAOYSA-D
SMILES: [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
Grade 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (9)
  • (3)
  • (3)