accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OCeO

CAS RN 1306-38-3

IUPAC Name: dioxocerium
Synonyms: cerium oxide
Molecular Weight (g/mol): 172.11
Molecular Formula: CeO2
InChi Key: CETPSERCERDGAM-UHFFFAOYSA-N
SMILES: O=[Ce]=O
Molecular Weight (g/mol) 
  • (1)
  • (34)
  • (3)
Physical Form 
  • (2)
  • (4)
  • (1)
  • (6)
  • (2)
  • (22)
  • (1)
Quantity 
  • (2)
  • (1)
  • (12)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (11)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (4)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (34)
  • (1)
  • (34)
  • (3)
  • (2)
  • (4)
  • (1)
  • (6)
  • (2)
  • (22)
  • (1)
  • (2)
  • (1)
  • (12)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (11)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)