accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13061-96-6

IUPAC Name: methylboronic acid
Synonyms: methylboronic acid
Molecular Weight (g/mol): 59.86
Molecular Formula: CH5BO2
InChi Key: KTMKRRPZPWUYKK-UHFFFAOYSA-N
SMILES: CB(O)O
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results