accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 130645-48-6

IUPAC Name: 2-bromo-4-methylpyrimidine
Synonyms: 2-bromo-4-methylpyrimidine
Molecular Weight (g/mol): 173.01
Molecular Formula: C5H5BrN2
InChi Key: UZIJEBOLOXOVFY-UHFFFAOYSA-N
SMILES: CC1=NC(Br)=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results