accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1307-96-6

OCo

CAS RN 1307-96-6

IUPAC Name: oxocobalt
Synonyms: cobalt(II) oxide
Molecular Weight (g/mol): 74.93
Molecular Formula: CoO
InChi Key: IVMYJDGYRUAWML-UHFFFAOYSA-N
SMILES: O=[Co]
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (7)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)