accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1307-99-9

CoSe

CAS RN 1307-99-9

IUPAC Name: selanylidenecobalt
Synonyms: cobalt(II) selenide
Molecular Weight (g/mol): 137.9
Molecular Formula: CoSe
InChi Key: QVYIMIJFGKEJDW-UHFFFAOYSA-N
SMILES: [Co]=[Se]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results